Search
Search

par-1| Gene Symbol: par-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028847.4 | NP_001024018.1 | 12 - 13 | 2976 | 155 | |
| NM_001028848.4 | NP_001024019.1 | 9 - 10 | 2704 | 155 | ||
| NM_001047680.4 | NP_001041145.1 | 9 - 10 | 2586 | 155 | ||
| NM_001129495.4 | NP_001122967.2 | 9 - 10 | 2784 | 155 | ||
| NM_001269630.3 | NP_001256559.1 | 9 - 10 | 2709 | 155 | ||
| NM_001269631.3 | NP_001256560.1 | 12 - 13 | 3000 | 155 | ||
| NM_001269632.3 | NP_001256561.1 | 8 - 9 | 2523 | 155 | ||
| NM_001269633.3 | NP_001256562.1 | 5 - 6 | 1515 | 155 | ||
| NM_001269634.3 | NP_001256563.1 | 4 - 5 | 1182 | 155 | ||
| NM_001307761.3 | NP_001294690.1 | 9 - 10 | 2985 | 155 | ||
| NM_001373155.1 | NP_001359744.1 | 10 - 11 | 3232 | 155 | ||
| GenBank mRNA | U40858.1 | - | 9 - 10 | 2704 | 155 | |
Set Membership: |
|