Search
Search

ncam-1| Gene Symbol: ncam-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171615.7 | NP_741707.3 | 11 - 12 | 3658 | 144 | |
| NM_171616.5 | NP_741708.1 | 11 - 12 | 2542 | 144 | ||
| NM_171617.5 | NP_741709.1 | 10 - 11 | 2682 | 144 | ||
| GenBank mRNA | AF456360.2 | - | 11 - 12 | 2566 | 144 | |
Set Membership: |
|