Search
Search

mec-2| Gene Symbol: mec-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029395.4 | NP_001024566.1 | 5 - 6 | 659 | 74 | |
| NM_001029396.4 | NP_001024567.1 | 4 - 5 | 698 | 74 | ||
| NM_001368504.4 | NP_001355460.1 | 1 | 413 | 74 | ||
| NM_001368505.3 | NP_001355457.1 | 4 - 5 | 482 | 74 | ||
| NM_001368507.2 | NP_001355459.1 | 4 - 5 | 482 | 74 | ||
| NM_001368508.5 | NP_001355458.1 | 5 - 6 | 721 | 74 | ||
| NM_001368511.3 | NP_001355478.1 | 4 - 5 | 458 | 74 | ||
| NM_001368514.4 | NP_001355480.1 | 5 | 515 | 74 | ||
| NM_001368515.4 | NP_001355479.1 | 4 | 434 | 74 | ||
| NM_001380971.2 | NP_001368710.1 | 4 - 5 | 599 | 74 | ||
| NM_001380972.1 | NP_001368712.1 | 5 - 6 | 659 | 74 | ||
| NM_001380973.1 | NP_001368715.1 | 5 - 6 | 659 | 74 | ||
| NM_001380974.1 | NP_001368713.1 | 4 - 5 | 458 | 74 | ||
| NM_001380975.1 | NP_001368716.1 | 4 - 5 | 458 | 74 | ||
| NM_001380976.1 | NP_001368714.1 | 4 - 5 | 434 | 74 | ||
| NM_001380977.1 | NP_001368717.1 | 4 - 5 | 434 | 74 | ||
| NM_171691.6 | NP_741797.1 | 5 - 6 | 702 | 74 | ||
| GenBank mRNA | U26735.1 | - | 5 - 6 | 696 | 74 | |
Set Membership: |
|