Search
Search

nhx-5| Gene Symbol: nhx-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029549.4 | NP_001024720.1 | 1 - 2 | 273 | 99 | |
| NM_001373485.2 | NP_001359642.1 | 1 - 2 | 273 | 99 | ||
| GenBank mRNA | AF497828.1 | - | 1 - 2 | 294 | 99 | |
| AF497829.1 | - | 1 - 2 | 294 | 99 | ||
Set Membership: |
|