Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: nhr-48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375186.2 | NP_001362159.1 | 12 - 13 | 2127 | 98 | |
NM_001375187.2 | NP_001362160.1 | 12 - 13 | 2094 | 98 | ||
NM_001375188.2 | NP_001361843.1 | 13 - 14 | 2242 | 98 | ||
NM_001375189.1 | NP_001361880.1 | 13 - 14 | 2209 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |