Search
Search

clp-1| Gene Symbol: clp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379816.2 | NP_001367451.1 | 4 - 5 | 433 | 109 | |
| NM_001379817.1 | NP_001367450.1 | 3 - 4 | 769 | 109 | ||
| NM_066339.6 | NP_498740.2 | 3 - 4 | 453 | 109 | ||
| NM_066340.6 | NP_498741.3 | 5 - 6 | 666 | 109 | ||
Set Membership: |
|