Search
Search

ajm-1| Gene Symbol: ajm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330906.2 | NP_001317734.1 | 13 - 14 | 1965 | 88 | |
| NM_001330908.3 | NP_001317889.1 | 7 - 8 | 1145 | 88 | ||
| NM_001330909.3 | NP_001317890.1 | 7 - 8 | 920 | 88 | ||
| NM_001330910.3 | NP_001317891.1 | 5 - 6 | 788 | 88 | ||
| NM_001330911.3 | NP_001317888.1 | 4 - 5 | 674 | 88 | ||
| NM_001330912.3 | NP_001317897.1 | 3 - 4 | 358 | 88 | ||
| NM_001330913.3 | NP_001317898.1 | 2 - 3 | 113 | 88 | ||
| NM_001381045.1 | NP_001368606.1 | 13 - 14 | 1928 | 88 | ||
| NM_001381046.2 | NP_001367383.1 | 13 - 14 | 1936 | 88 | ||
| NM_001381047.2 | NP_001367382.1 | 13 - 14 | 1928 | 88 | ||
| NM_001381048.1 | NP_001367381.1 | 13 - 14 | 1965 | 88 | ||
Set Membership: |
|