Search
Search

nhr-80| Gene Symbol: nhr-80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064725.7 | NP_497126.1 | 3 - 4 | 561 | 68 | |
| NM_181901.2 | NP_871630.1 | 2 - 3 | 499 | 68 | ||
| GenBank mRNA | AY204179.1 | - | 3 - 4 | 489 | 68 | |
Set Membership: |
|