Search
Search

pqn-35| Gene Symbol: pqn-35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381396.1 | NP_001367814.1 | 4 - 5 | 813 | 83 | |
| NM_001381397.2 | NP_001367813.1 | 5 - 6 | 885 | 83 | ||
Set Membership: |
|