Search
Search

pde-4| Gene Symbol: pde-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047333.6 | NP_001040798.1 | 2 - 3 | 271 | 64 | |
| NM_001306543.3 | NP_001293472.1 | 2 - 3 | 124 | 64 | ||
| NM_001383892.1 | 2 - 3 | 184 | 64 | |||
| NM_063199.6 | NP_495600.1 | 2 - 3 | 206 | 64 | ||
Set Membership: |
|