Search
Search

| Gene Symbol: crn-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381579.1 | NP_001367040.1 | 3 - 4 | 2170 | 80 | |
| NM_001383937.1 | 5 - 6 | 3489 | 80 | |||
| GenBank mRNA | AY303577.1 | - | 3 - 4 | 2188 | 80 | |
| Gene Symbol: algn-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063881.6 | NP_496282.2 | 8 - 9 | 4318 | 80 | |
Set Membership: |
|