Search
Search

lntl-1| Gene Symbol: lntl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380241.1 | NP_001367493.1 | 4 - 5 | 583 | 75 | |
| NM_001380242.1 | NP_001368410.1 | 1 - 2 | 121 | 75 | ||
| NM_068465.6 | NP_500866.2 | 5 - 6 | 761 | 75 | ||
| NR_131524.1 | 4 - 5 | 583 | 75 | |||
Set Membership: |
|