Search
Search

lin-40| Gene Symbol: lin-40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330804.3 | NP_001317834.1 | 1 - 2 | 1845 | 68 | |
| NM_001373016.2 | NP_001360139.1 | 3 - 4 | 2274 | 68 | ||
| NM_071630.5 | NP_504031.1 | 3 - 4 | 2267 | 68 | ||
| NM_071631.7 | NP_504032.1 | 3 - 4 | 2274 | 68 | ||
Set Membership: |
|