Search
Search

dys-1| Gene Symbol: dys-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306310.3 | NP_001293239.1 | 17 - 18 | 3733 | 58 | |
| NM_001306311.3 | NP_001293240.1 | 6 - 7 | 934 | 58 | ||
| NM_001306313.4 | NP_001293242.1 | 20 - 21 | 4378 | 58 | ||
| NM_001306314.3 | NP_001293243.1 | 16 - 17 | 3700 | 58 | ||
| NM_001383731.1 | 16 - 17 | 3751 | 58 | |||
| NM_060545.6 | NP_492946.1 | 21 - 22 | 4473 | 58 | ||
Set Membership: |
|