Search
Search

scrm-1| Gene Symbol: scrm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001264776.4 | NP_001251705.1 | 2 - 3 | 668 | 69 | |
| NM_001373645.1 | NP_001359603.1 | 2 - 3 | 662 | 69 | ||
| NM_060575.5 | NP_492976.2 | 2 - 3 | 662 | 69 | ||
Set Membership: |
|