Search
Search

| Gene Symbol: gpi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026667.7 | NP_001021838.1 | 4 - 5 | 1501 | 76 | |
| NM_001383747.1 | 4 - 5 | 1338 | 76 | |||
| Gene Symbol: ubc-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383748.1 | 4 - 5 | 1355 | 76 | ||
Set Membership: |
|