Search
Search

lev-11| Gene Symbol: lev-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026524.7 | NP_001021695.1 | 8 - 9 | 1066 | 76 | |
| NM_001313524.3 | NP_001300453.1 | 6 - 7 | 687 | 76 | ||
| NM_001313525.4 | NP_001300454.1 | 6 - 7 | 699 | 76 | ||
| NM_001313526.3 | NP_001300455.1 | 5 - 6 | 498 | 76 | ||
| NM_001313527.3 | NP_001300456.1 | 4 | 384 | 76 | ||
| NM_001313528.4 | NP_001300457.1 | 6 | 825 | 76 | ||
| NM_001313529.3 | NP_001300458.1 | 5 | 771 | 76 | ||
| NM_001313530.3 | NP_001300459.1 | 5 | 771 | 76 | ||
| NM_001313531.3 | NP_001300460.1 | 6 | 826 | 76 | ||
| NM_001313532.4 | NP_001300461.1 | 5 - 6 | 789 | 76 | ||
| NM_001381266.2 | NP_001367087.1 | 8 - 9 | 679 | 76 | ||
| GenBank mRNA | D38540.1 | - | 8 - 9 | 835 | 76 | |
Set Membership: |
|