Search
Search

cwn-1| Gene Symbol: cwn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061267.5 | NP_493668.1 | 5 - 6 | 663 | 90 | |
| NR_132572.1 | 5 - 6 | 642 | 90 | |||
| GenBank mRNA | X72941.1 | - | 657 | 90 | ||
Set Membership: |
|