Search
Search

nhl-3| Gene Symbol: nhl-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047352.4 | NP_001040817.2 | 8 - 9 | 1203 | 93 | |
| NM_001047353.6 | NP_001040818.2 | 5 - 6 | 903 | 93 | ||
| NM_001316864.3 | NP_001303793.1 | 2 - 3 | 444 | 93 | ||
Set Membership: |
|