Search
Search

mpk-2| Gene Symbol: mpk-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026742.5 | NP_001021913.1 | 5 - 6 | 788 | 96 | |
| NM_062545.8 | NP_494946.2 | 5 - 6 | 767 | 96 | ||
| NM_062546.8 | NP_494947.2 | 5 - 6 | 756 | 96 | ||
Set Membership: |
|