Search
Search

clr-1| Gene Symbol: clr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027037.4 | NP_001022208.1 | 16 - 17 | 3847 | 77 | |
| NM_001027038.3 | NP_001022209.1 | 17 - 18 | 3933 | 77 | ||
| NM_001136345.4 | NP_001129817.1 | 16 - 17 | 3842 | 77 | ||
| NM_001347284.3 | NP_001334206.1 | 17 - 18 | 3798 | 77 | ||
| NR_144560.1 | 16 - 17 | 3725 | 77 | |||
Set Membership: |
|