Search
Search

tat-4| Gene Symbol: tat-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383859.1 | 15 - 16 | 3633 | 104 | ||
| NR_165322.1 | 15 - 16 | 3579 | 104 | |||
| NR_165323.1 | 15 - 16 | 3579 | 104 | |||
Set Membership: |
|