Search
Search

spp-11| Gene Symbol: spp-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001365320.2 | NP_001352251.1 | 6 - 7 | 457 | 87 | |
| NM_001368591.4 | NP_001355419.1 | 5 | 855 | 87 | ||
| NM_001368592.3 | NP_001355420.1 | 6 | 420 | 87 | ||
Set Membership: |
|