Search
Search

glr-4| Gene Symbol: glr-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267197.2 | NP_001254126.1 | 10 - 11 | 1402 | 110 | |
| NM_001373747.3 | NP_001359784.1 | 10 - 11 | 1405 | 110 | ||
Set Membership: |
|