Search
Search

ltd-1| Gene Symbol: ltd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322746.3 | NP_001309573.1 | 4 - 5 | 779 | 141 | |
| NM_063296.6 | NP_495697.2 | 7 - 8 | 1546 | 141 | ||
| GenBank mRNA | AF489839.1 | - | 7 - 8 | 1568 | 141 | |
Set Membership: |
|