Search
Search

slc-25A32| Gene Symbol: slc-25A32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129147.5 | NP_001122619.1 | 1 - 2 | 148 | 89 | |
| NM_063345.6 | NP_495746.1 | 1 - 2 | 204 | 89 | ||
Set Membership: |
|