Search
Search

| Gene Symbol: jun-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027195.4 | NP_001022366.1 | 7 - 8 | 957 | 68 | |
| NM_001027196.5 | NP_001022367.1 | 7 - 8 | 822 | 68 | ||
| NM_001027197.5 | NP_001022368.1 | 2 - 3 | 247 | 68 | ||
| NM_001136355.4 | NP_001129827.1 | 2 - 3 | 379 | 68 | ||
| NM_001356878.2 | NP_001343785.1 | 6 - 7 | 820 | 68 | ||
| NM_001356879.3 | NP_001343787.1 | 5 - 6 | 625 | 68 | ||
| NM_001356880.3 | NP_001343788.1 | 4 - 5 | 514 | 68 | ||
| NM_001356881.3 | NP_001343786.1 | 3 - 4 | 391 | 68 | ||
| NM_001383910.1 | 8 - 9 | 1139 | 68 | |||
| GenBank mRNA | EU553920.1 | - | 7 - 8 | 895 | 68 | |
| EU553922.1 | - | 5 - 6 | 661 | 68 | ||
| EU553923.1 | - | 2 - 3 | 331 | 68 | ||
| EU553925.1 | - | 2 - 3 | 220 | 68 | ||
| Gene Symbol: C16D2.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381554.1 | NP_001367159.1 | 7 - 8 | 980 | 68 | |
| NM_063547.5 | NP_495948.1 | 2 - 3 | 474 | 68 | ||
Set Membership: |
|