Search
Search

| Gene Symbol: sym-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063559.9 | NP_495960.2 | 2 - 3 | 327 | 63 | |
| Gene Symbol: twk-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001356885.2 | NP_001343566.1 | 4 | 639 | 63 | |
Set Membership: |
|