Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: let-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136358.4 | NP_001129830.1 | 3 - 4 | 527 | 146 | |
NM_001136359.5 | NP_001129831.1 | 5 - 6 | 725 | 146 | ||
NM_001313678.3 | NP_001300607.1 | 6 - 7 | 752 | 146 | ||
NM_063561.5 | NP_495962.2 | 5 - 6 | 799 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |