Search
Search

mel-11| Gene Symbol: mel-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026757.4 | NP_001021928.1 | 11 - 12 | 2620 | 97 | |
| NM_001381562.1 | NP_001367038.1 | 10 - 11 | 2310 | 97 | ||
| NM_063593.6 | NP_495994.1 | 10 - 11 | 2302 | 97 | ||
| GenBank mRNA | U86640.1 | - | 10 - 11 | 2293 | 97 | |
Set Membership: |
|