Search
Search

ufd-3| Gene Symbol: ufd-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267303.3 | NP_001254232.1 | 2 - 3 | 413 | 79 | |
| NM_063745.3 | NP_496146.1 | 12 - 13 | 3101 | 79 | ||
| NM_063746.5 | NP_496147.1 | 8 - 9 | 2368 | 79 | ||
Set Membership: |
|