Search
Search

clpp-1| Gene Symbol: clpp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267310.4 | NP_001254239.1 | 3 - 4 | 466 | 131 | |
| NM_001267311.3 | NP_001254240.1 | 3 - 4 | 415 | 131 | ||
| NM_001267312.3 | NP_001254241.1 | 1 - 2 | 139 | 131 | ||
Set Membership: |
|