Search
Search

tag-180| Gene Symbol: tag-180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027192.5 | NP_001022363.1 | 16 - 17 | 2973 | 78 | |
| NM_001027193.5 | NP_001022364.1 | 15 - 16 | 3217 | 78 | ||
Set Membership: |
|