Search
Search

kel-1| Gene Symbol: kel-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267381.2 | NP_001254310.1 | 8 - 9 | 1828 | 62 | |
| NM_001267382.3 | NP_001254311.1 | 3 - 4 | 286 | 62 | ||
| GenBank mRNA | AB023424.1 | - | 8 - 9 | 1820 | 62 | |
Set Membership: |
|