Search
Search

gpb-1| Gene Symbol: gpb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381616.2 | NP_001366714.1 | 7 - 8 | 700 | 60 | |
| NM_001383961.1 | 8 - 9 | 951 | 60 | |||
| GenBank mRNA | AF291846.1 | - | 7 - 8 | 697 | 60 | |
Set Membership: |
|