Search
Search

unc-52| Gene Symbol: unc-52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027317.8 | NP_001022488.1 | 21 - 22 | 7006 | 97 | |
| NM_001267510.3 | NP_001254439.1 | 22 - 23 | 6417 | 97 | ||
| NM_001267511.3 | NP_001254440.1 | 20 - 21 | 6636 | 97 | ||
| NM_001267513.3 | NP_001254442.1 | 20 - 21 | 6427 | 97 | ||
| NM_001267514.4 | NP_001254443.1 | 21 - 22 | 6615 | 97 | ||
| NM_001267515.3 | NP_001254444.1 | 22 - 23 | 6897 | 97 | ||
| NM_001267516.3 | NP_001254445.1 | 22 - 23 | 6912 | 97 | ||
| NM_001267517.3 | NP_001254446.1 | 23 - 24 | 7185 | 97 | ||
| NM_001306656.3 | NP_001293585.1 | 1 - 2 | 159 | 97 | ||
| NM_001350388.3 | NP_001337294.1 | 32 - 33 | 9561 | 97 | ||
| NM_001377905.1 | NP_001364664.1 | 32 - 33 | 9454 | 97 | ||
| NM_064643.10 | NP_497044.3 | 23 - 24 | 7194 | 97 | ||
| NM_064644.6 | NP_497045.1 | 21 - 22 | 6723 | 97 | ||
| NM_064645.6 | NP_497046.1 | 23 - 24 | 7285 | 97 | ||
Set Membership: |
|