Search
Search

pde-3| Gene Symbol: pde-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267523.2 | NP_001254452.1 | 9 - 10 | 1800 | 72 | |
| NM_001267525.3 | NP_001254454.1 | 8 - 9 | 1478 | 72 | ||
| NM_001306640.4 | NP_001293569.1 | 10 - 11 | 1795 | 72 | ||
| NM_001306642.3 | NP_001293571.1 | 12 - 13 | 2036 | 72 | ||
| NM_001306643.3 | NP_001293572.1 | 11 - 12 | 1877 | 72 | ||
| NM_064683.6 | NP_497084.2 | 7 - 8 | 1586 | 72 | ||
Set Membership: |
|