Search
Search

| Gene Symbol: K02F3.12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027485.3 | NP_001022656.1 | 4 - 5 | 1519 | 92 | |
| NM_001027486.5 | NP_001022657.1 | 4 - 5 | 1440 | 92 | ||
| Gene Symbol: exos-4.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381715.2 | NP_001367942.1 | 4 - 5 | 1442 | 92 | |
| Gene Symbol: tag-51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064876.9 | NP_497277.2 | 7 - 8 | 3213 | 92 | |
Set Membership: |
|