Search
Search

trp-2| Gene Symbol: trp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027532.5 | NP_001022703.1 | 9 - 10 | 1425 | 66 | |
| NM_064910.8 | NP_497311.2 | 15 - 16 | 2684 | 66 | ||
| GenBank mRNA | AJ276028.1 | - | 14 - 15 | 2514 | 66 | |
Set Membership: |
|