Search
Search

sup-26| Gene Symbol: sup-26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267963.3 | NP_001254892.1 | 2 - 3 | 441 | 70 | |
| NM_001267969.1 | NP_001254898.1 | 2 - 3 | 441 | 70 | ||
| NM_001267971.4 | NP_001254900.1 | 3 - 4 | 348 | 70 | ||
| NM_001381827.1 | NP_001367246.1 | 2 - 3 | 240 | 70 | ||
| NM_001381830.2 | NP_001366727.1 | 3 - 4 | 557 | 70 | ||
Set Membership: |
|