Search
Search

cku-80| Gene Symbol: cku-80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322514.2 | NP_001309438.1 | 12 - 13 | 2059 | 79 | |
| NM_001322520.3 | NP_001309599.1 | 5 - 6 | 908 | 79 | ||
| NM_001322524.3 | NP_001309600.1 | 3 - 4 | 593 | 79 | ||
Set Membership: |
|