Search
Search

sel-5| Gene Symbol: sel-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027391.8 | NP_001022562.1 | 7 - 8 | 1000 | 147 | |
| NM_001027392.6 | NP_001022563.1 | 7 - 8 | 1000 | 147 | ||
| GenBank mRNA | AF256466.1 | - | 7 - 8 | 990 | 147 | |
| AF256467.1 | - | 7 - 8 | 990 | 147 | ||
Set Membership: |
|