Search
Search

nca-2| Gene Symbol: nca-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136363.4 | NP_001129835.1 | 4 - 5 | 993 | 147 | |
| NM_065653.6 | NP_498054.2 | 5 - 6 | 1149 | 147 | ||
| GenBank mRNA | AY555272.2 | - | 5 - 6 | 1149 | 147 | |
| DQ917241.1 | - | 4 - 5 | 1031 | 147 | ||
Set Membership: |
|