Search
Search

larp-1| Gene Symbol: larp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047402.3 | NP_001040867.2 | 3 - 4 | 373 | 74 | |
| NM_001047403.5 | NP_001040868.3 | 6 - 7 | 1192 | 74 | ||
Set Membership: |
|