Search
Search

par-3| Gene Symbol: par-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027436.3 | NP_001022607.1 | 13 - 14 | 4220 | 58 | |
| NM_001306735.2 | NP_001293664.1 | 10 - 11 | 4075 | 58 | ||
| NM_001306736.3 | NP_001293665.1 | 10 - 11 | 4066 | 58 | ||
| NM_001306739.3 | NP_001293668.1 | 14 - 15 | 4384 | 58 | ||
| NM_001306740.3 | NP_001293669.1 | 14 - 15 | 4393 | 58 | ||
| NM_001306741.3 | NP_001293670.1 | 12 - 13 | 4064 | 58 | ||
| NM_001372593.2 | NP_001360234.1 | 9 - 10 | 3893 | 58 | ||
| NM_001372595.2 | NP_001360237.1 | 8 - 9 | 3746 | 58 | ||
| NM_001379720.2 | NP_001367341.1 | 9 - 10 | 4022 | 58 | ||
| NM_001382875.1 | 13 - 14 | 4207 | 58 | |||
| GenBank mRNA | U25032.1 | - | 13 - 14 | 4219 | 58 | |
Set Membership: |
|