Search
Search

rpn-6.1| Gene Symbol: rpn-6.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027450.4 | NP_001022621.1 | 2 - 3 | 368 | 145 | |
| NM_001027451.5 | NP_001022622.1 | 3 - 4 | 349 | 145 | ||
Set Membership: |
|