Search
Search

fbn-1| Gene Symbol: fbn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276780.3 | NP_001263709.1 | 10 - 11 | 4172 | 59 | |
| NM_001276782.3 | NP_001263711.1 | 10 - 11 | 4176 | 59 | ||
| NM_001276783.3 | NP_001263712.1 | 10 - 11 | 4166 | 59 | ||
| NM_001276785.3 | NP_001263714.1 | 10 - 11 | 4166 | 59 | ||
| NM_001276786.3 | NP_001263715.1 | 10 - 11 | 4166 | 59 | ||
| NM_001276787.4 | NP_001263716.1 | 10 - 11 | 4166 | 59 | ||
| NM_001276788.3 | NP_001263717.1 | 10 - 11 | 4166 | 59 | ||
| NM_001276789.3 | NP_001263718.1 | 10 - 11 | 4166 | 59 | ||
| NM_066269.6 | NP_498670.2 | 10 - 11 | 4166 | 59 | ||
| GenBank mRNA | JQ990141.1 | - | 10 - 11 | 4172 | 59 | |
Set Membership: |
|