Search
Search

fmo-3| Gene Symbol: fmo-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001351929.3 | NP_001338838.1 | 6 - 7 | 1241 | 67 | |
| NM_066955.6 | NP_499356.1 | 6 - 7 | 1332 | 67 | ||
| GenBank mRNA | AJ582072.1 | - | 6 - 7 | 1251 | 67 | |
Set Membership: |
|