Search
Search

taf-12| Gene Symbol: taf-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268215.3 | NP_001255144.1 | 2 - 3 | 235 | 66 | |
| NM_001268216.3 | NP_001255145.1 | 1 - 2 | 132 | 66 | ||
Set Membership: |
|