Search
Search

cua-1| Gene Symbol: cua-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268273.3 | NP_001255202.1 | 7 - 8 | 1140 | 121 | |
| NM_001268274.3 | NP_001255203.1 | 4 - 5 | 641 | 121 | ||
| GenBank mRNA | D83665.1 | - | 6 - 7 | 1076 | 121 | |
Set Membership: |
|